1,10-Phenanthroline monohydrate 99+%, pure

Supplier: Thermo Scientific Chemicals
Danger

13013-0050 13013-0250 13013-1000 13013-5000
TS13013-0050EA 55 USD
TS13013-0050 TS13013-0250 TS13013-1000 TS13013-5000
1,10-Phenanthroline monohydrate 99+%, pure
1,10-Phenanthroline monohydrate
Formula: C₁₂H₈N₂·1H₂O
Melting Pt: 97…101 °C
Storage Temperature: Ambient
MDL Number: MFCD00149973
CAS Number: 5144-89-8
EINECS: 200-629-2
UN: 2811
ADR: 6.1,III
Merck Index: 1307291
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aba346b9f67fdda9afd4d48b893e6318564d604c&vid=2d08e04ee9525293a747b55cdcff8f66a6c4715a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to cream Crystalline powder or needles
Infrared spectrum Conforms
Melting point 98 to 102 °C
Titration with HClO4 ≥99.0 %
UV at 260 nm A: ≤0.02
Titration after Ion exchange ≥99.0 % (on dry substance)
Membrane filtration test (on 0.5 M soln.) Passes test
Suitability (HPLC gradient) Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR