2-Nitrophenol 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
12874-0010
12874-0050
12874-1000
12874-5000
TS12874-0010EA
105
USD
TS12874-0010
TS12874-0050
TS12874-1000
TS12874-5000
2-Nitrophenol 99%
2-Nitrophenol
Formula:
C₆H₅NO₃ Boiling Pt: 214.6 °C (1013 hPa) Melting Pt: 43…47 °C Flash Pt: 108 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011688 CAS Number: 88-75-5 EINECS: 201-857-5 UN: 1663 ADR: 6.1,III Merck Index: 1206717 |
Specification Test Results
Appearance | Yellow to yellow-green Powder or crystals and/or chunks or low melting crystalline mass |
Infrared spectrum | Conforms |
Melting point | 43 to 47 °C |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...