Girard's reagent T (carbazoylmethyltrimethylammonium chloride) 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Hydrazinyl-N,N,N-trimethyl-2-oxoethanaminium chloride

11987-0500 11987-2500
TS11987-0500EA 41 USD
TS11987-0500 TS11987-2500
Girard's reagent T (carbazoylmethyltrimethylammonium chloride) 99%
Girard's reagent T (Carbazoylmethyltrimethylammonium chloride)
Formula: C₅H₁₄ClN₃O
MW: 167.64 g/mol
Melting Pt: 188…192 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012009
CAS Number: 123-46-6
EINECS: 204-629-3

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 187 to 192 °C
Titration Argentometric ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR