(1-Butyl)triphenylphosphonium bromide 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Butyltriphenylphosphonium bromide
10806-0050
10806-1000
TS10806-0050EA
24
USD
TS10806-0050
TS10806-1000
(1-Butyl)triphenylphosphonium bromide 99%
(1-Butyl)triphenylphosphonium bromide
Formula:
H₃C(CH₂)₃P(C₆H₅)₃Br Melting Pt: 240…243 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011855 CAS Number: 1779-51-7 EINECS: 217-219-4 UN: 3464 ADR: 6.1,III |
|
Specification Test Results
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Melting point | 238 to 243 °C |
Titration Argentometric | ≥98.5 % |
Specific optical rotation | -9.5° to -12.5° (20 °C, 589 nm) (c=1, methanol) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...