Tris EDTA buffer, for molecular biology, DNAse, RNAse, Protease free ready to use, pH 8.0

Supplier: Thermo Scientific
32734-5000
TS32734-5000EA 133.71 USD
TS32734-5000
Tris EDTA buffer, for molecular biology, DNAse, RNAse, Protease free ready to use, pH 8.0
Buffers Biological Buffers
CAS No.: 38641-82-6

MDL No.: MFCD00236359
Appearance: Clear colourless solution
Storage temperature: Ambient
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR