Sand 40-100 mesh, pure

Supplier: Thermo Scientific Chemicals

Synonyms: Silicium dioxide, Silicium (IV) oxide, Silicon dioxide

37094-0010 37094-0050 37094-0250 370942500
TS37094-0010EA 62 USD
TS37094-0010 TS37094-0050 TS37094-0250 TS370942500
Sand 40-100 mesh, pure
Sand
Formula: SiO₂
MW: 60.08
Boiling Pt: 2230 °C
Melting Pt: 1700
Density: 2.6
Storage Temperature: Ambient
MDL Number: MFCD02100519
CAS Number: 7631-86-9
EINECS: 231-545-4

Order Now

Specification Test Results

Appearance Light beige to gray Crystals
Additional info Cr2O3: 0.00005 %
Particle size 205 µm (D50) Typical


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR