β-Nicotinamide adenine dinucleotide hydrate 98+%

Supplier: Thermo Scientific Chemicals

12453-0050 12453-0010 12453-2500
TS12453-0050EA 346 USD
TS12453-0050 TS12453-0010 TS12453-2500
β-Nicotinamide adenine dinucleotide hydrate 98+%
β-Nicotinamide adenine dinucleotide hydrate
Formula: C₂₁H₂₇N₇O₁₄P₂.xH₂O
Storage Temperature: Freezer
MDL Number: MFCD00150377
CAS Number: 53-84-9
EINECS: 200-184-4
Merck Index: 15,6429

Order Now

Specification Test Results

Appearance White to yellow Powder
Infrared spectrum Conforms
Water ≤11 % (K.F.)
Solubility (1 % in water) Clear colorless
UV spectrophotometry ≥98.0 %
Molar absorption coefficient ≥17000 l/(mol.cm) (Lambda max 260 nm, pH 7.5)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR