tri-Calcium phosphate 96%, pure

Supplier: Thermo Scientific Chemicals

Synonyms: Calcium phosphate, Tricalcium orthophosphate, penta-Calcium hydroxide triphosphate, Calcium phosphate tribasic

44639-0010 44639-0050
TS44639-0010EA 135 USD
TS44639-0010 TS44639-0050
tri-Calcium phosphate 96%, pure
tri-Calcium phosphate
Formula: Ca₃(PO₄)₂
Melting Pt: 1670 °C
Storage Temperature: Ambient
MDL Number: MFCD00015984
CAS Number: 7758-87-4

Order Now

Specification Test Results

Appearance White Powder
Titration Complexometric ≥95 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR