Zinc chloride 99.99% (trace metals basis), extra pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Zinc dichloride, Butter of zinc, Dichlorozinc, Zinc (II) chloride

31817-0500 AC31817-0100
TS31817-0500EA 340 USD
TS31817-0500 TS31817-0100
Zinc chloride 99.99% (trace metals basis), extra pure
Zinc chloride
Formula: ZnCl₂
MW: 136.3 g/mol
Boiling Pt: 732 °C (760 mmHg)
Melting Pt: 293 °C
Storage Temperature: Ambient
MDL Number: MFCD00011295
CAS Number: 7646-85-7
EINECS: 231-592-0
UN: 2331
ADR: 8,III
Merck Index: 13,10185

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals and/or chunks
Total trace metal impurities ≤0.01 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR