Charcoal activated, NORIT® GAC 1240 12-40 mesh

Supplier: Thermo Scientific Chemicals

Synonyms: Activated carbonCarbon activatedActivated charcoal

39571-2500 39571-0010
TS39571-0010EA 201.28 USD
TS39571-0010 TS39571-2500
Charcoal activated, NORIT® GAC 1240 12-40 mesh
Charcoal activated
Formula: C
MW: 12.01 g/mol
Melting Pt: 3652 °C
Storage Temperature: Ambient
MDL Number: MFCD00133992
CAS Number: 7440-44-0

Order Now

Specification Test Results

Appearance Black Granular solid
Water ≤2 % (As packed)
Particle size ≤5 % (> 12 mesh)
Particle size ≤4 % (< 40 mesh)
Molasses number ≥230
Abrasion number ≥75
Iodine number ≥1000 mg/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR