Linezolid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Zyvox, (S)-N-((3-(3-fluoro-4-morpholinophenyl)-2-oxooxazolidin-5-yl)methyl)acetamide

46059-0010 46059-2500
TS46059-0010EA 421 USD
TS46059-0010 TS46059-2500
Linezolid 98%
Linezolid
Formula: C₁₆H₂₀FN₃O₄
MW: 337.4 g/mol
Melting Pt: 181…183 °C
Storage Temperature: Ambient
MDL Number: MFCD00937825
CAS Number: 165800-03-3

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Loss on drying ≤0.5 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR