β-(N-Morpholino)ethanesulfonic acid (MES) hydrate 99+% for biochemistry

Supplier: Thermo Scientific Chemicals

17259-0025 17259-0250 17259-1000 17259-5000
TS17259-0025EA 1898 USD
TS17259-0025 TS17259-0250 TS17259-1000 TS17259-5000
β-(N-Morpholino)ethanesulfonic acid (MES) hydrate 99+% for biochemistry
β-(N-Morpholino)ethanesulfonic acid (MES) hydrate
Formula: C₆H₁₃NO₄S·xH₂O
Melting Pt: 243 °C
Storage Temperature: Ambient
MDL Number: MFCD00149409
CAS Number: 1266615-59-1
EINECS: 224-632-3
Merck Index: 13,05929

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Water ≤9.0 % (K.F.)
Solubility (5 % w/v in water) Clear colorless
UV at 260 nm A: ≤0.05
pH 2.5 to 4.0 (1 % in water at 25°C)
Titration with NaOH ≥99.0 % (On dry substance)
Chloride (Cl) ≤100 ppm
Sulfate (SO4) ≤500 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR