MBD 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 7-(4-Methoxybenzylamino)-4-nitrobenzoxadiazole

41354-0010 41354-0050
TS41354-0010EA 198 USD
TS41354-0010 TS41354-0050
MBD 98%
MBD
Formula: C₁₄H₁₂N₄O₄
MW: 300.274 g/mol
Melting Pt: 176…180 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00042778
CAS Number: 33984-50-8

Order Now

Specification Test Results

Appearance Orange to brown Powder
Infrared spectrum Conforms
Melting point 177 to 182 °C
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR