N-Succinimidyl Acrylate 99%

Supplier: Thermo Scientific Chemicals
Warning

40030-0050 40030-0010 40030-0250
TS40030-0050EA 259 USD
TS40030-0050 TS40030-0010 TS40030-0250
N-Succinimidyl Acrylate 99%
N-Succinimidyl Acrylate
Formula: C₇H₇NO₄
Melting Pt: 69 °C
Storage Temperature: Freezer
MDL Number: MFCD00078261
CAS Number: 38862-24-7

Order Now

Specification Test Results

Appearance White to yellow Powder or crystals and/or chunks
Infrared spectrum Conforms
Titration with TBAH ≥98.5 %
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR