Di(N-succinimidyl) carbonate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
20960-0050
20960-0250
TS20960-0050EA
140
USD
TS20960-0050
TS20960-0250
Di(N-succinimidyl) carbonate 98%
Di(N-succinimidyl) carbonate
Formula:
C₉H₈N₂O₇ Melting Pt: 190 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00009767 CAS Number: 74124-79-1 |
Specification Test Results
Appearance | White to light yellow Crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...