Rose Bengal 85%, pure, high purity biological stain

Supplier: Thermo Scientific Chemicals

18945-1000 18945-0010 18945-0050 18945-0250 18945-5000
TS18945-1000EA 228 USD
TS18945-1000 TS18945-0010 TS18945-0050 TS18945-0250 TS18945-5000
Rose Bengal 85%, pure, high purity biological stain
Rose Bengal
Formula: C₂₀H₂Cl₄I₄O₅Na₂
Storage Temperature: Ambient
MDL Number: MFCD00151169
CAS Number: 632-69-9

Order Now

Specification Test Results

Appearance Bordeaux to red-brown Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤10 % (105°C)
Dye content ≥80 %
UV Lambda max 540 to 550 nm (in water)
Absorption Ratio P-15/P+15 1.21 to 1.95


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR