5-Aminolevulinic acid hydrochloride 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
5-Amino-4-ketovaleric acid hydrochloride, 5-Amino-4-oxopentanoic acid hydrochloride, δ-Aminolevulinic Acid Hydrochloride
10392-0050
10392-5000
TS10392-0050EA
520
USD
TS10392-0050
TS10392-5000
5-Aminolevulinic acid hydrochloride 98+%
5-Aminolevulinic acid hydrochloride
Formula:
C₅H₁₀ClNO₃ Melting Pt: 156…158 °C (dec.) Storage Temperature: Freezer |
MDL Number:
MFCD00012869 CAS Number: 5451-09-2 |
Specification Test Results
Appearance | White to light yellow Crystalline powder or crystals |
Infrared spectrum | Conforms |
Titration Argentometric | ≥98.0 to 102.0 % |
Loss on drying | ≤2.0 % (105°C, 3 h) (vacuum) |
Sulfated ash | ≤0.5 % |
Solubility | (5 % in water) Clear to slightly hazy colorless to light yellow |
HPLC | ≥98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...