Isosorbide dimethyl ether 99%

Supplier: Thermo Scientific Chemicals

Synonyms: (3R,3aR,6S,6aR)-3,6-Dimethoxyhexahydrofuro[3,2-b]furan

36991-1000 36991-0250
TS36991-1000EA 215 USD
TS36991-1000 TS36991-0250
Isosorbide dimethyl ether 99%
Isosorbide dimethyl ether
Formula: C₈H₁₄O₄
Boiling Pt: 93…95 °C (.1hPa)
Flash Pt: 120 °C
Storage Temperature: Ambient
MDL Number: MFCD00064828
CAS Number: 5306-85-4
EINECS: 226-159-8

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4600 to 1.4620 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR