Tetrabutylammonium dihydrogen phosphate 99% for HPLC
Supplier: Thermo Scientific Chemicals
Synonyms:
Tetra-n-butylammonium phosphate monobasic, Tetrabutylammonium phosphate monobasic, Tetra-n-butylammonium dihydrogen phosphate, IPC-TBA-P, Tetra-n-butylammonium phosphate
42016-0250
42016-1000
AC42016-0050
TS42016-0250EA
1147
USD
TS42016-0250
TS42016-1000
TS42016-0050
Tetrabutylammonium dihydrogen phosphate 99% for HPLC
Tetrabutylammonium dihydrogen phosphate
Formula:
C₁₆H₃₈NO₄P MW: 339.46 g/mol Melting Pt: 148…151 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064526 CAS Number: 5574-97-0 |
Specification Test Results
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
UV | at 214 nm A: ≤0.05 |
Titration with NaOH | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...