Azidotrimethylsilane 94%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Trimethylsilyl azide

16793-2500 AC16793-0100 AC16793-0500
TS16793-2500EA 817 USD
TS16793-2500 TS16793-0100 TS16793-0500
Azidotrimethylsilane 94%
Azidotrimethylsilane
Formula: C₃H₉N₃Si
MW: 115.21 g/mol
Boiling Pt: 95…96 °C (760 mmHg)
Density: 0.876 g/cm³
Flash Pt: 6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001986
CAS Number: 4648-54-8
EINECS: 225-078-5
UN: 1992
ADR: 3 II

Order Now

Specification Test Results

Appearance Clear to slightly hazy colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4110 to 1.4180 (20°C, 589 nm)
GC ≥92 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR