N,N'-Bis(3-aminopropyl)propane-1,3-diamine 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
N,N'-Bis(3-aminopropyl)-1,3-propanediamine, 1,5,9,13-Tetraazatridecane
40228-0250
40228-0050
TS40228-0250EA
568
USD
TS40228-0250
TS40228-0050
N,N'-Bis(3-aminopropyl)propane-1,3-diamine 97%
N,N'-Bis(3-aminopropyl)propane-1,3-diamine
Formula:
C₉H₂₄N₄ Boiling Pt: 102 °C (1 mmHg) Flash Pt: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008213 CAS Number: 4605-14-5 EINECS: 225-007-8 UN: 2735 ADR: 8,II |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4900 to 1.4930 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...