tert-Butyllithium 1.9 M in pentane, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: t-Butyllithium

39654-1000 AC39654-8000
TS39654-1000EA 102 USD
TS39654-1000 TS39654-8000
tert-Butyllithium 1.9 M in pentane, AcroSeal™
tert-Butyllithium
Formula: C₄H₉Li
Boiling Pt: 36…38 °C (760 mmHg)
Density: 0.69 g/cm³
Flash Pt: –56 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008795
CAS Number: 594-19-4
UN: 3394
ADR: 4.2,

Order Now

Specification Test Results

Appearance Clear to hazy colorless to yellow Solution (May develop some turbidity or precipitate)
Titration Active base 1.80 to 2.02 M


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR