TES (2-(Tris(hydroxymethyl)methylamino)ethane-1-sulfonic acid) 99% Biochemistry

Supplier: Thermo Scientific Chemicals

Synonyms: TES Buffer, N-Tris(hydroxymethyl)methyl-2-aminoethanesulfonic acid, N-Tris(hydroxymethyl)methyl-2-aminoethanesulphonic acid

20751-1000 20751-0250
TS20751-1000EA 183 USD
TS20751-1000 TS20751-0250
TES (2-(Tris(hydroxymethyl)methylamino)ethane-1-sulfonic acid) 99% Biochemistry
TES (2-(Tris(hydroxymethyl)methylamino)ethane-1-sulphonic acid)
Formula: C₆H₁₅NO₆S
MW: 229.25 g/mol
Melting Pt: 223…225 °C
Storage Temperature: Ambient
MDL Number: MFCD00007532
CAS Number: 7365-44-8
EINECS: 230-906-3

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤1 %
Solubility (0.1 M in water) Clear colorless
UV at 280 nm A: ≤0.02
pH 4 to 8 (0.1 M at 20°C)
Titration with NaOH ≥98.5 %
Heavy metals (as Pb) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR