p-Toluenesulfonyl isocyanate 96%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Tosyl isocyanate, 4-Isocyanatosulphonyltoluene, 4-Isocyanatosulfonyltoluene

18307-0250 18307-5000 18307-1000
TS18307-0250EA 51 USD
TS18307-0250 TS18307-5000 TS18307-1000
p-Toluenesulfonyl isocyanate 96%
p-Toluenesulphonyl isocyanate
Formula: C₈H₇NO₃S
MW: 197.21 g/mol
Boiling Pt: 144 °C (10 mmHg)
Melting Pt: –2 °C
Density: 1.29 g/cm³
Flash Pt: 145 °C
Storage Temperature: Ambient
MDL Number: MFCD00002030
CAS Number: 4083-64-1
EINECS: 223-810-8
UN: 2206
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5330 to 1.5360 (20°C, 589 nm)
GC ≥95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR