Acrylic acid 98% stabilised, extra pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Prop-2-enoic acid

16425-0010 16425-0025 16425-0100 16425-2500
TS16425-0010EA 126 USD
TS16425-0010 TS16425-0025 TS16425-0100 TS16425-2500
Acrylic acid 98% stabilised, extra pure
Acrylic acid
Formula: C₃H₄O₂
MW: 72.06 g/mol
Boiling Pt: 141.6
Melting Pt: 13
Density: 1.05 g/cm³
Flash Pt: 48 °C
Storage Temperature: Ambient
MDL Number: MFCD00004367
CAS Number: 79-10-7
EINECS: 201-177-9
UN: 2218
ADR: 8 II

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4190 to 1.4230 (20°C, 589 nm)
Water ≤0.1 % (K.F.)
GC ≥97.5 %
Color scale ≤20 APHA
Stabilizer 180 to 220 ppm MEHQ
Dimer ≤2.5 % diacrylic acid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR