Resazurin sodium salt, pure, certified
Supplier: Thermo Scientific Chemicals
|
Warning
|
41890-0050
41890-0250
41890-0010
TS41890-0050EA
84
USD
TS41890-0050
TS41890-0250
TS41890-0010
Resazurin sodium salt, pure, certified
Resazurin sodium salt
Formula:
C₁₂H₆NNaO₄ Storage Temperature: Ambient |
MDL Number:
MFCD00005036 CAS Number: 62758-13-8 |
Specification Test Results
Appearance | Dark green to black Powder |
Infrared spectrum | Conforms |
UV spectrum | Complies |
Dye content | ≥80 % |
BSC Certification | Passes |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...