Methacrylic acid 99.5% stabilised, extra pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Methylpropenoic acid

16831-0040 16831-0251 16831-0250 168310010
TS16831-0040EA 163 USD
TS16831-0040 TS16831-0251 TS16831-0010 TS16831-0250 TS16831-2500 TS16831-5000 TS168310010
Methacrylic acid 99.5% stabilised, extra pure
Methacrylic acid
Formula: C₄H₆O₂
MW: 86.09 g/mol
Boiling Pt: 163
Melting Pt: 15
Density: 1.01 g/cm³
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00002651
CAS Number: 79-41-4
EINECS: 201-204-4
UN: 2531
ADR: 8 II
Merck Index: 13,05967

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4295 to 1.4315 (20°C, 589 nm)
Water ≤0.2 %
GC ≥99.4 %
Color scale ≤20 APHA
Stabilizer ca. 250 ppm MEHQ


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR