(±)-2-Methyltetrahydrofuran 99+% stabilised, pure

Supplier: Thermo Scientific Chemicals
Danger

16836-1000 168360025 168360100 168365000 168360010
TS16836-1000EA 73 USD
TS16836-1000 TS168360025 TS168360100 TS168365000 TS168360010
(±)-2-Methyltetrahydrofuran 99+% stabilised, pure
(±)-2-Methyltetrahydrofuran
Formula: C₅H₁₀O
MW: 86.13 g/mol
Boiling Pt: 79
Melting Pt: -136
Density: 0.86 g/cm³
Flash Pt: –11 °C
Storage Temperature: Ambient
MDL Number: MFCD00005367
CAS Number: 96-47-9
EINECS: 202-507-4
UN: 2536
ADR: 3 II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4046 to 1.4066 (20°C, 589 nm)
Water ≤0.2 % (K.F.)
GC ≥99.0 %
Stabilizer 150 to 400 ppm BHT


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR