tert-Butylchlorodimethylsilane 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: tert-Butyldimethylsilyl Chloride, Chloro-tert-butyldimethylsilane, t-Butyldimethylchlorosilane, tert-Butyldimethylchlorosilane, tert-Butyldimethylsilyl chloride, TBDMS chloride

18393-1000 18393-5000 AC18393-0050 AC18393-0250
TS18393-1000EA 307 USD
TS18393-1000 TS18393-5000 TS18393-0050 TS18393-0250
tert-Butylchlorodimethylsilane 98%
tert-Butylchlorodimethylsilane
Formula: C₆H₁₅ClSi
Boiling Pt: 125 °C (1013 hPa)
Melting Pt: 85…91 °C
Density: 0.87 g/cm³
Flash Pt: 22 °C
MDL Number: MFCD00000501
CAS Number: 18162-48-6
EINECS: 242-042-4
UN: 2921

Order Now

Specification Test Results

Appearance Colorless to white Crystals
Infrared spectrum Conforms
Melting point 85 to 91 °C
GC ≥97.5 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR