PIPES 98.5+% for biochemistry

Supplier: Thermo Scientific Chemicals

Synonyms: Piperazine-1,4-bis(2-ethanesulphonic acid), Piperazine-1,4-bis(2-ethanesulfonic acid), 2,2'-(Piperazine-1,4-diyl)diethanesulphonic acid, 2,2'-(Piperazine-1,4-diyl)diethanesulfonic acid, Piperazine-N,N'-bis-2-ethanesulphonic acid, 2,2'-(Piperazine-1,4-diyl)bis(ethanesulfonic) acid, 2,2'-(Piperazine-1,4-diyl)bis(ethanesulphonic) acid, Piperazine-N,N'-bis-2-ethanesulfonic acid

17261-5000 17261-0025 17261-0250 17261-1000
TS17261-5000EA 408 USD
TS17261-5000 TS17261-0025 TS17261-0250 TS17261-1000
PIPES 98.5+% for biochemistry
PIPES
Formula: C₈H₁₈N₂O₆S₂
MW: 302.37 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00006159
CAS Number: 5625-37-6
EINECS: 227-057-6
Merck Index: 12,07633
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5192468b680c7bea85a31a131811618356a531b9&vid=ef0e3d488c68ea7f6f583fede6ebf7d2196392b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤0.5 %
UV at 280 nm A: ≤0.04
Titration with NaOH ≥98.50 %
Iron (Fe) ≤15 ppm
Lead (Pb) ≤5 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR