Platinum(II) chloride (≥73% Pt)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Platinum dichloride

19538-0010 AC19538-0050 AC19538-0250 AC19538-2500
TS19538-0010EA 200 USD
TS19538-0010 TS19538-0050 TS19538-0250 TS19538-2500
Platinum(II) chloride (≥73% Pt)
Platinum (II) chloride
Formula: Cl₂Pt
MW: 265.99 g/mol
Melting Pt: 581 °C (dec.)
Density: 6.05 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011181
CAS Number: 10025-65-7
EINECS: 233-034-1
UN: 3260
ADR: 8,III
Merck Index: 12,07687

Order Now

Specification Test Results

Appearance Gray-green to brown Powder
Assay ≥71.8 % (Pt)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR