4,7,13,16,21,24-Hexaoxa-1,10-diazabicyclo[8.8.8]hexacosane (Kryptofix® 222) 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Cryptand 2.2.2, Fludeoxyglucose Related Compound A, Kryptofix 222, 4,7,13,16,21,24-Hexaoxa-1,10-diazabicyclo(8.8.8)hexacosane, 4,7,13,16,21,24-Hexaoxa-1,10-diazabicyclo[8.8.8]hexacosane, Cryptand 222, (2.2.2)Cryptand

29195-0010 29195-0050 29195-2500
TS29195-0010EA 202 USD
TS29195-0010 TS29195-0050 TS29195-2500
4,7,13,16,21,24-Hexaoxa-1,10-diazabicyclo[8.8.8]hexacosane (Kryptofix® 222) 98%
4,7,13,16,21,24-Hexaoxa-1,10-diazabicyclo[8.8.8]hexacosane (Kryptofix® 222)
Formula: C₁₈H₃₆N₂O₆
MW: 376.49 g/mol
Melting Pt: 68…72 °C
Storage Temperature: Ambient
MDL Number: MFCD00005111
CAS Number: 23978-09-8
EINECS: 245-962-4
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a74ac532561e4fecf111f3260b402bcd1fb0af9&vid=13491901da172b5971e6d7f330c893460a77cd23&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 68 to 72 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR