Trifluoromethanesulfonic acid 98%, Extra Pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Trifluoromethanesulphonic acid, Trifluoromethanesulfonic acid

16989-0100 16989-0500 16989-0010 16989-0011 16989-2500
TS16989-0100EA 80 USD
TS16989-0100 TS16989-0500 TS16989-0010 TS16989-0011 TS16989-2500
Trifluoromethanesulfonic acid 98%, Extra Pure
Trifluoromethanesulphonic acid
Formula: CHF₃O₃S
MW: 150.08 g/mol
Boiling Pt: 57…60 °C (760 mmHg)
Melting Pt: –131 °C
Density: 0.82 g/cm³
Flash Pt: –34 °C
Storage Temperature: Ambient
MDL Number: MFCD00007514
CAS Number: 1493-13-6
EINECS: 200-861-4
UN: 2402
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4240 to 1.4270 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR