Triphenylphosphine 99%

Supplier: Thermo Scientific Chemicals
Warning

14042-2500 14042-0010 14042-0050 14042-0250
TS14042-2500EA 121 USD
TS14042-2500 TS14042-0010 TS14042-0050 TS14042-0250
Triphenylphosphine 99%
Triphenylphosphine
Formula: C₁₈H₁₅P
Boiling Pt: 377 °C (760 mmHg)
Melting Pt: 78.5…81.5 °C
Flash Pt: 181 °C
Storage Temperature: Ambient
MDL Number: MFCD00003043
CAS Number: 603-35-0
EINECS: 210-036-0
Merck Index: 12,09873

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals or flakes
Infrared spectrum Conforms
Melting point 78.5 to 81.5 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR