Tetramethylsilane 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: TMS

26886-1000 26886-5000 AC26886-0025
TS26886-1000EA 117 USD
TS26886-1000 TS26886-5000 TS26886-0025
Tetramethylsilane 99%
Tetramethylsilane
Formula: C₄H₁₂Si
MW: 88.22 g/mol
Boiling Pt: 26.6 °C (1013 hPa)
Melting Pt: –99 °C
Density: 0.64 g/cm³
Flash Pt: –27 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008274
CAS Number: 75-76-3
EINECS: 200-899-1
UN: 2749
ADR: 3,I

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3575 to 1.3595 (20°C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR