Magnesium sulfate heptahydrate 98+% ACS
Supplier: Thermo Scientific Chemicals
Synonyms:
sulphuric acid magnesium salt heptahydrate, sulfuric acid magnesium salt heptahydrate, Epsom salt, Bitter salt
42390-0010
AC42390-0050
AC42390-0250
AC42390-5000
TS42390-0010EA
147
USD
TS42390-0010
TS42390-0050
TS42390-0250
TS42390-5000
Magnesium sulfate heptahydrate 98+% ACS
Magnesium sulphate heptahydrate
Formula:
MgSO₄·7H₂O MW: 246.48 g/mol Melting Pt: 770 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149785 CAS Number: 10034-99-8 EINECS: 231-298-2 Merck Index: 13,05715 |
Specification Test Results
Appearance | White Crystals |
Titration Complexometric | 98.0 to 102.0 % |
pH | 5.0 to 8.2 (5% soln. at 25°C) |
Chloride (Cl) | ≤5 ppm |
Nitrate (NO3) | ≤0.002 % |
Ammonium (NH4) | ≤0.002 % |
Calcium (Ca) | ≤0.02 % |
Iron (Fe) | ≤5 ppm |
Manganese (Mn) | ≤5 ppm |
Potassium (K) | ≤0.005 % |
Sodium (Na) | ≤0.005 % |
Strontium (Sr) | ≤0.005 % |
Heavy metals (ICP-OES) | ≤5 ppm |
Insoluble matter | ≤0.005 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...