Diisopropyl ether 99+% stabilised, extra pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Isopropyl ether

Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

18068-0025 180680250
TS18068-0025EA 184 USD
TS18068-0025 TS180680010 TS180680250
Diisopropyl ether 99+% stabilised, extra pure
Diisopropyl ether
Formula: (CH₃)₂CHOCH(CH₃)₂
Boiling Pt: 68.5 °C (1013 hPa)
Melting Pt: –85.5 °C
Density: 0.72 g/cm³
Flash Pt: –29 °C
Storage Temperature: Ambient
MDL Number: MFCD00008880
CAS Number: 108-20-3
UN: 1159
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=932b8c021b2bfa9a11b8d8812cafbafecc6f9239&vid=dea267e876203eb2c2674787701ec602da1b3cf6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3670 to 1.3690 (20°C, 589 nm)
Water ≤0.1 %
GC ≥99.0 %
Color scale ≤10 APHA
Stabilizer 5 to 250 ppm BHT
Residue after evaporation ≤0.01 %
Acidity ≤0.005 %
Peroxides ≤0.002 % (as H2O2)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR