Diisopropylamine 99%

Supplier: Thermo Scientific Chemicals
Danger

14946-0025 14946-0010
TS14946-0025EA 161 USD
TS14946-0025 TS14946-0010
Diisopropylamine 99%
Diisopropylamine
Formula: C₆H₁₅N
Boiling Pt: 84 °C (1013 hPa)
Melting Pt: –61 °C
Density: 0.716 g/cm³
Flash Pt: –17 °C
Storage Temperature: Ambient
MDL Number: MFCD00008862
CAS Number: 108-18-9
EINECS: 203-558-5
UN: 1158
ADR: 3,II
Merck Index: 13,03219

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3910 to 1.3930 (20°C, 589 nm)
GC ≥98.5 %
Titration with NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR