Propylene oxide 99%, pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (±)-Methyloxirane, (±)-Propene oxide, (±)-Propylene oxide

14962-0025 14962-0010
TS14962-0025EA 119 USD
TS14962-0025 TS14962-0010
Propylene oxide 99%, pure
Propylene oxide
Formula: C₃H₆O
MW: 58.08 g/mol
Boiling Pt: 34.3
Melting Pt: -112
Density: 0.83 g/cm³
Flash Pt: –37 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005126
CAS Number: 75-56-9
EINECS: 200-879-2
UN: 1280
ADR: 3 I
Merck Index: 12,08041
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=443494ff0620e18a4c5de7f3f4cd87d6e5eec44c&vid=33bc95a664261fbc904256d1b87a1972dde5dcd1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3650 to 1.3670 (20°C, 589 nm)
Water ≤0.02 %
GC ≥98.5 %
Color scale ≤10 APHA
Aldehyde (C3H6O) ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR