1,4-Cyclohexadiene 99+% stabilised
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Cyclohexa-1,4-diene
15482-1000
15482-0050
15482-0250
TS15482-1000EA
424
USD
TS15482-1000
TS15482-0050
TS15482-0250
1,4-Cyclohexadiene 99+% stabilised
1,4-Cyclohexadiene
Formula:
C₆H₈ MW: 80.13 g/mol Boiling Pt: 82 Melting Pt: -49.2 Density: 0.83 g/cm³ Flash Pt: –45 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001535 CAS Number: 628-41-1 EINECS: 200-837-3 UN: 2363 ADR: 3 I Merck Index: 12,03771 |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4296 to 1.4316 (20°C, 589 nm) |
GC | ≥99.0 % |
Cloud point | ≤20.0°C |
Calcium (Ca) | ≤30 ppm |
Iron (Fe) | ≤10 ppm |
Sodium (Na) | ≤200 ppm |
Insoluble matter | ≤0.005 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...