Hydroxylammonium chloride 99+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Hydroxylamine HCl, Hydroxylamine hydrochloride

27010-0010 27010-0050 27010-1000 27010-2500
TS27010-0010EA 174 USD
TS27010-0010 TS27010-0050 TS27010-1000 TS27010-2500
Hydroxylammonium chloride 99+%
Hydroxylammonium chloride
Formula: NH₂OH·HCl
MW: 69.49 g/mol
Boiling Pt: 306
Melting Pt: 151 (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00051089
CAS Number: 5470-11-1
EINECS: 226-798-2
UN: 2923
ADR: 8 III
Merck Index: 13,04853

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Titration with NaOH 99.0 to 101.0 %
Sulfate (SO4) ≤0.005 %
Iron (Fe) ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR