Barium hydroxide octahydrate 98+% ACS

Supplier: Thermo Scientific Chemicals
Danger

42346-1000 AC42346-0025 AC42346-5000
TS42346-1000EA 44 USD
TS42346-1000 TS42346-0025 TS42346-5000
Barium hydroxide octahydrate 98+% ACS
Barium hydroxide octahydrate
Formula: Ba(OH)₂·8H₂O
Boiling Pt: 780 °C (1013 hPa)
Melting Pt: 78 °C
Density: 2.18 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00149152
CAS Number: 12230-71-6
EINECS: 241-234-5
UN: 3262
ADR: 8,II
Merck Index: 13,00980

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals or flakes
Titration with HCl ≥98.0 %
Chloride (Cl) ≤0.001 %
Sulfide (S) (Limit about 0.001 %) Passes test
Carbonate (as BaCO3) ≤2.0 %
Calcium (Ca) ≤0.05 %
Iron (Fe) ≤0.001 %
Potassium (K) ≤0.01 %
Sodium (Na) ≤0.01 %
Strontium (Sr) ≤0.8 %
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.01 % (in dilute HCl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR