Dimethylamine 2 M in THF, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

38838-1000 38838-8000
TS38838-1000EA 84 USD
TS38838-1000 TS38838-8000
Dimethylamine 2 M in THF, AcroSeal™
Dimethylamine
Formula: C₂H₇N
MW: 45.08
Boiling Pt: 6.9
Melting Pt: -92.8
Density: 0.85 g/cm³
Flash Pt: –36 °C
Storage Temperature: Ambient
MDL Number: MFCD00008288
CAS Number: 124-40-3
EINECS: 204-697-4
UN: 2924
ADR: 3 II
Merck Index: 12,03278

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Solution
Strength 1.8 to 2.2 M


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR