Boron tribromide 1 M in methylene chloride
Supplier: Thermo Scientific Chemicals
|
Danger
|
19890-1000
19890-8000
198900100
TS19890-1000EA
201
USD
TS19890-1000
TS19890-8000
TS198900100
Boron tribromide 1 M in methylene chloride
Boron tribromide
Formula:
BBr₃ MW: 250.52 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011312 CAS Number: 10294-33-4 EINECS: 233-657-9 UN: 3390 ADR: 6.1, |
Specification Test Results
Appearance | Clear yellow to dark brown Solution (May darken on storage. May develop some turbidity or precipitate) |
Strength | 0.95 to 1.10 M |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...