Boron tribromide 99+%

Supplier: Thermo Scientific Chemicals
Danger

36521-1000 AC36521-0250
TS36521-1000EA 119 USD
TS36521-1000 TS36521-0250
Boron tribromide 99+%
Boron tribromide
Formula: BBr₃
MW: 250.52 g/mol
Boiling Pt: 91.2 °C (760 mmHg)
Melting Pt: –46 °C
Density: 2.65 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011312
CAS Number: 10294-33-4
EINECS: 233-657-9
UN: 2692
ADR: 8,I
Merck Index: 13,01337

Order Now

Specification Test Results

Appearance Clear amber Liquid
Titration ≥99.0 %
Sulfur (S) ≤100 ppm
Heavy metals (as Pb) ≤100 ppm
Impurity ≤0.2 % Free bromine (Br2)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR