Hydrobromic acid ∼48% in water ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Hydrogen bromide

42378-5000 423780025
TS42378-5000EA 141 USD
TS42378-5000 TS423780025
Hydrobromic acid ∼48% in water ACS
Hydrogen bromide
Formula: HBr
MW: 80.91 g/mol
Melting Pt: –11 °C
Density: 1.48 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011323
CAS Number: 10035-10-6
UN: 1788
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Solution
Residue after ignition ≤0.002 %
Titration with NaOH 47.0 to 49.0 %
Chloride (Cl) ≤500 ppm
Iodide (I) ≤30 ppm
Phosphate (PO4) ≤10 ppm
SO4 and SO3 (as SO4) ≤0.003 %
Iron (Fe) ≤1 ppm
Selenium (Se) ≤0.01 ppm
Heavy metals (ICP-OES) ≤5 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR