Naphthacene 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,3-Benzanthracene, Benz[b]anthracene, Tetracene

10526-0050 10526-0010 10526-5000
TS10526-0050EA 773 USD
TS10526-0050 TS10526-0010 TS10526-5000
Naphthacene 98%
Naphthacene
Formula: C₁₈H₁₂
MW: 228.29 g/mol
Boiling Pt: 437
Melting Pt: 356
Storage Temperature: Ambient
MDL Number: MFCD00003702
CAS Number: 92-24-0
EINECS: 202-138-9
UN: 3077
ADR: 9 III
Merck Index: 12,06456

Order Now

Specification Test Results

Appearance Orange Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR