Imidazole ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Glyoxaline, 1H-Imidazole, Glyoxalamine

39674-5000 39674-0025 39674-1000
TS39674-5000EA 177 USD
TS39674-5000 TS39674-0025 TS39674-1000
Imidazole ACS
Imidazole
Formula: C₃H₄N₂
MW: 68.08 g/mol
Boiling Pt: 255…256 °C (760 mmHg)
Melting Pt: 86…90 °C
Flash Pt: 145 °C
Storage Temperature: Ambient
MDL Number: MFCD00005183
CAS Number: 288-32-4
UN: 3263
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=48eba49e61201d6fffa2bef0c54633a20660c7ef&vid=e7f154e4976b52732539e49bbff60454d5eb82ff&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to yellow Powder or crystals
Infrared spectrum Conforms
Water ≤0.2 %
Residue after ignition ≤0.1 %
pH 9.5 to 11.0 (5% at 25°C)
Titration with HClO4 ≥99 %
Iron (Fe) ≤0.001 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR