Decahydronaphthalene (mixture of cis and trans isomers) cis- and trans- mixture 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Decalin, Bicyclo[4.4.0]decane, Decahydronaphthalene

11184-0025 111840250 111845000 111840010
TS11184-0025EA 277 USD
TS11184-0025 TS111840250 TS111845000 TS111840010
Decahydronaphthalene (mixture of cis and trans isomers) cis- and trans- mixture 98%
Decahydronaphthalene (mixture of cis and trans isomers)
Formula: C₁₀H₁₈
MW: 138.25 g/mol
Boiling Pt: 187 °C (760 mmHg)
Melting Pt: -40
Density: 0.89 g/cm³
Flash Pt: 58 °C
Storage Temperature: Ambient
MDL Number: MFCD00004130
CAS Number: 91-17-8
EINECS: 202-046-9
UN: 1147
ADR: 3 III
Merck Index: 13,02866
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=41b5fff8aa81be0e42af177a8f0e3ac149943c6e&vid=ca9f3c5d71ac621a6216f8b8b28d7cb27b29fd84&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4730 to 1.4760 (20°C, 589 nm)
Water ≤0.1 %
GC ≥97.5 %
Color scale ≤10 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR