Sulfur 99.999% (trace metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
19993-0500
19993-2500
199930100
TS19993-0500EA
112
USD
TS19993-0500
TS19993-2500
TS199930100
Sulfur 99.999% (trace metals basis)
Sulphur
Formula:
S Boiling Pt: 445 °C (1013 hPa) Melting Pt: 114 °C Flash Pt: 168 °C Storage Temperature: Ambient |
MDL Number:
MFCD00085316 CAS Number: 7704-34-9 EINECS: 231-722-6 UN: 1350 ADR: 4.1,III Merck Index: 13,09059 |
Specification Test Results
Appearance | Yellow Pieces |
Particle size | ≤12 mm |
Total trace metal impurities | ≤0.001 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...